Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021391.3 | NP_067366.1 | 6 | 584 | ||
GenBank mRNA | AF281676.1 | - | 6 | 474 | ||
AK002813.1 | - | 2 | 218 | |||
BC025123.1 | - | 6 | 553 | 4019442 | ||
CA321941.1 | - | 6 | 584 |