Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021393.2 | NP_067368.2 | 4 | 734 | ||
GenBank mRNA | AB041607.1 | - | 3 | 855 | ||
AK046129.1 | - | 1 | 1399 | |||
AK080419.1 | - | 4 | 436 | |||
AK087524.1 | - | 4 | 434 | |||
AK173124.1 | - | 3 | 269 | |||
AK184385.1 | - | 1 | 34 | |||
AK198680.1 | - | 3 | 241 | |||
AK203337.1 | - | 2 | 116 | |||
AK217288.1 | - | 1 | 72 | |||
BC004762.1 | - | 4 | 622 | 3155904 | ||
BU519080.1 | - | 4 | 667 |