Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021296.2 | NP_067271.1 | 4 | 1927 | ||
GenBank mRNA | AK082983.1 | - | 1918 | |||
AK145493.1 | - | 4 | 1919 | |||
BC024866.1 | - | 1 | 336 | |||
BC068232.1 | - | 4 | 1904 | 30533079 | ||
BC094583.1 | - | 4 | 1894 | |||
BC120747.1 | - | 4 | 1917 | 40129670 |