Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021311.3 | NP_067286.1 | 22 | 2958 | ||
GenBank mRNA | AB032604.1 | - | 22 | 2923 | ||
AF438405.1 | - | 22 | 2959 | |||
BC129857.1 | - | 21 | 2808 | 40109791 | ||
BC129858.1 | - | 20 | 2912 | 40109793 |