Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271476.1 | NP_001258405.1 | 5, 6 | 1809 | ||
NM_001271477.1 | NP_001258406.1 | 4, 5 | 1672 | |||
NM_001271479.1 | NP_001258408.1 | 3, 4 | 1363 | |||
NM_021319.5 | NP_067294.2 | 4, 5 | 1701 | |||
GenBank mRNA | AB084140.1 | - | 5, 6 | 1806 | ||
AB084141.1 | - | 5, 6 | 1719 | |||
AF149837.1 | - | 4, 5 | 1698 | |||
AF149838.1 | - | 4, 5 | 1611 | |||
AF315578.1 | - | 3, 4 | 1360 | |||
AF315584.1 | - | 4, 5 | 1571 | |||
AF317548.1 | - | 5, 6 | 1567 | |||
AK050019.1 | - | 4, 5 | 1673 | |||
AY282722.1 | - | 4, 5 | 1639 | |||
BC019396.1 | - | 4, 5 | 1674 | 5124925 |