Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021339.2 | NP_067314.2 | 13 | 2606 | ||
GenBank mRNA | AF090866.1 | - | 13 | 2606 | ||
BC109176.2 | - | 13 | 2605 | 40048829 | ||
CV557123.1 | - | 2 | 219 |