Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rqcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021383.5 | NP_067358.1 | 2113 | |||
GenBank mRNA | AK005025.2 | - | 8 | 2115 | ||
AK011335.1 | - | 1 | 539 | |||
BC034663.1 | - | 1 | 1119 | |||
BC050898.1 | - | 8 | 1914 | 6401907 | ||
BC051948.1 | - | 1953 | 5708192 |