Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167730.1 | NP_001161202.1 | 5 | 476 | ||
NM_021385.2 | NP_067360.2 | 5 | 476 | |||
GenBank mRNA | AF205278.1 | - | 5 | 400 | ||
AK012795.1 | - | 5 | 477 | |||
AK028505.1 | - | 5 | 396 | |||
AK030043.1 | - | 5 | 477 | |||
AK048975.1 | - | 5 | 451 | |||
BC011120.1 | - | 5 | 439 | 4014013 |