Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021390.3 | NP_067365.2 | 3 | 1926 | ||
GenBank mRNA | AB051409.1 | - | 2 | 1780 | ||
BC062937.1 | - | 3 | 1926 | 6839137 |