Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyou1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021395.4 | NP_067370.3 | 26 | 4363 | ||
GenBank mRNA | AK128937.1 | - | 26 | 4306 | ||
AK145857.1 | - | 26 | 4312 | |||
BC050107.1 | - | 25 | 4270 | 30012694 |