Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020781.3 | NP_065832.1 | 3 | 560 | ||
NM_170686.2 | NP_733787.1 | 2 | 497 | |||
XM_011516439.2 | XP_011514741.1 | 3 | 378 | |||
XM_011516440.2 | XP_011514742.1 | 2 | 350 | |||
XM_011516441.2 | XP_011514743.1 | 2 | 333 | |||
GenBank mRNA | AK026380.1 | - | 2 | 498 | ||
AK290499.1 | - | 2 | 465 | |||
AK301890.1 | - | 3 | 378 | |||
AY049743.1 | - | 3 | 560 | |||
AY049744.1 | - | 2 | 239 | |||
BC043295.2 | - | 2 | 489 | 5298104 |