Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021411.4 | NP_067386.3 | 1, 2 | 122 | ||
GenBank mRNA | AB232633.1 | - | 1, 2 | 101 | ||
AF233582.1 | - | 1, 2 | 101 | |||
AK039716.1 | - | 1, 2 | 116 | |||
AK040216.1 | - | 1, 2 | 122 | |||
AK050483.1 | - | 1, 2 | 116 | |||
BC050757.1 | - | 1, 2 | 122 | |||
BC128020.1 | - | 1, 2 | 120 | 40136290 | ||
BY131048.1 | - | 1, 2 | 133 |