Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012449.2 | NP_036581.1 | 3 | 483 | ||
GenBank mRNA | AF186249.1 | - | 3 | 348 | ||
AK304076.1 | - | 3 | 382 | |||
AK308452.1 | - | 3 | 402 | |||
BC011802.2 | - | 3 | 387 | 4422866 |
Gene Symbol: STEAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164460.1 | NP_001157932.1 | 3 | 483 | ||
NM_207342.2 | NP_997225.1 | 3 | 426 | |||
GenBank mRNA | AK308991.1 | - | 3 | 380 | ||
BC066301.1 | - | 3 | 323 | 5274319 |