Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021435.3 | NP_067410.1 | 2 | 347 | ||
GenBank mRNA | AB041549.1 | - | 2 | 348 | ||
AK029046.1 | - | 2 | 264 | |||
AK029969.1 | - | 2 | 405 | |||
AK033913.1 | - | 2 | 277 | |||
AK042150.1 | - | 2 | 247 | |||
AK045791.1 | - | 2 | 289 | |||
AK138591.1 | - | 2 | 288 | |||
BC016275.1 | - | 2 | 278 | |||
BC033512.1 | - | 2 | 272 | 4017336 |