Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700123O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021437.2 | NP_067412.1 | 2 | 1076 | ||
GenBank mRNA | AB041657.1 | - | 2 | 1031 | ||
AK007261.1 | - | 2 | 477 | |||
AK167989.1 | - | 2 | 260 | |||
AK170890.1 | - | 2 | 409 | |||
BC011283.1 | - | 2 | 205 | 3581864 |