Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021452.1 | NP_067427.1 | 1 | 268 | ||
GenBank mRNA | AF215892.1 | - | 1 | 268 | ||
AK013647.1 | - | 1 | 627 | |||
AK050596.1 | - | 1 | 168 | |||
BC048471.1 | - | 1 | 281 | |||
BC117779.2 | - | 1 | 268 | 40091866 |