Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013943.2 | NP_039234.1 | 2 | 412 | ||
GenBank mRNA | AF097330.1 | - | 2 | 267 | ||
AF109196.1 | - | 2 | 205 | |||
AK001624.1 | - | 2 | 299 | |||
AK301466.1 | - | 2 | 210 | |||
AK315051.1 | - | 2 | 249 | |||
AL117424.1 | - | 2 | 324 | |||
BC012444.1 | - | 2 | 277 | 3861372 | ||
CR533501.1 | - | 2 | 127 | |||
DA904028.1 | - | 2 | 368 | |||
DB091421.1 | - | 2 | 415 |