Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021457.3 | NP_067432.2 | 1 | 1541 | ||
GenBank mRNA | AF054623.2 | - | 1 | 905 | ||
AK143101.1 | - | 1 | 1361 | |||
BC053010.1 | - | 1 | 1296 | 5697795 |
Set Membership: |
G-protein coupled receptor |