Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077712.2 | NP_001071180.1 | 5, 6 | 763 | ||
NM_001290713.1 | NP_001277642.1 | 4, 5 | 489 | |||
NM_021465.4 | NP_067440.3 | 5, 6 | 623 | |||
GenBank mRNA | AJ002636.1 | - | 4, 5 | 465 | ||
AK142460.1 | - | 5, 6 | 733 | |||
AK146718.1 | - | 5, 6 | 623 | |||
AK168899.1 | - | 5, 6 | 623 | |||
BC066041.1 | - | 5, 6 | 504 | |||
CO434048.1 | - | 3, 4 | 310 |