Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021473.3 | NP_067448.1 | 2, 3 | 434 | ||
GenBank mRNA | AF225564.1 | - | 2, 3 | 339 | ||
AK005376.1 | - | 2, 3 | 370 | |||
AK009462.1 | - | 2, 3 | 211 | |||
AK011321.1 | - | 2, 3 | 363 | |||
AK011388.1 | - | 2, 3 | 389 | |||
AK011667.1 | - | 2, 3 | 390 | |||
AK011794.1 | - | 2, 3 | 363 | |||
AK011856.1 | - | 2, 3 | 390 | |||
AK011906.1 | - | 2, 3 | 368 | |||
AK011908.1 | - | 2, 3 | 403 | |||
AK011918.1 | - | 2, 3 | 389 | |||
AK128906.1 | - | 2, 3 | 156 | |||
AK144198.1 | - | 2, 3 | 390 | |||
AK146081.1 | - | 2, 3 | 388 | |||
AK146278.1 | - | 2, 3 | 390 | |||
AK152086.1 | - | 2, 3 | 369 | |||
AK159573.1 | - | 2, 3 | 388 | |||
AK160185.1 | - | 2, 3 | 364 | |||
AK164461.1 | - | 2, 3 | 389 | |||
AK168950.1 | - | 2, 3 | 385 | |||
AK186333.1 | - | 2, 3 | 191 | |||
AK192030.1 | - | 2, 3 | 192 | |||
AK210646.1 | - | 2, 3 | 192 | |||
AK219186.1 | - | 2, 3 | 191 | |||
AY207463.1 | - | 2, 3 | 411 | |||
BC003850.1 | - | 2, 3 | 218 | |||
BC039926.1 | - | 2, 3 | 383 | 3156658 | ||
BC046762.1 | - | 2, 3 | 340 | 5693736 |