Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164352.1 | NP_001157824.1 | 3 | 539 | ||
NM_021474.3 | NP_067449.3 | 4 | 298 | |||
GenBank mRNA | AF104223.1 | - | 3 | 253 | ||
AF109122.1 | - | 3 | 512 | |||
AK075585.1 | - | 4 | 287 | |||
BC012269.1 | - | 3 | 436 | 3980048 | ||
BY234446.1 | - | 4 | 298 |