Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163028.1 | NP_001156500.1 | 3 | 372 | ||
NM_021486.3 | NP_067461.2 | 3 | 372 | |||
GenBank mRNA | AF271298.1 | - | 3 | 403 | ||
AF294899.1 | - | 3 | 385 | |||
AJ278064.2 | - | 3 | 384 | |||
AK054171.1 | - | 2 | 124 | |||
AK146472.1 | - | 3 | 369 | |||
BC125328.1 | - | 3 | 239 | 40129843 | ||
BC125330.1 | - | 3 | 239 | 40129845 | ||
BC145036.1 | - | 3 | 239 |