Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021493.2 | NP_067468.2 | 19 | 4315 | ||
GenBank mRNA | AB041572.1 | - | 5 | 1350 | ||
AK079135.1 | - | 3 | 1278 | |||
AK173187.1 | - | 10 | 1961 |