Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021510.2 | NP_067485.1 | 3 | 256 | ||
GenBank mRNA | AK052983.1 | - | 3 | 210 | ||
AK088169.1 | - | 3 | 209 | |||
AK161491.1 | - | 354 | ||||
AK219260.1 | - | 2 | 62 | |||
BC016459.1 | - | 3 | 224 | |||
BC027002.1 | - | 2 | 169 | |||
BC042187.1 | - | 3 | 191 | 5376052 | ||
BC056224.1 | - | 2 | 186 | 6515270 | ||
BY787002.1 | - | 3 | 256 | |||
Y14196.1 | - | 3 | 214 |