Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021434.5 | NP_067409.2 | 4 | 659 | ||
GenBank mRNA | AB041545.1 | - | 4 | 651 | ||
AK053422.1 | - | 4 | 629 | |||
AK169908.1 | - | 4 | 658 | |||
AK170316.1 | - | 4 | 624 | |||
AK181071.1 | - | 1 | 19 | |||
BC055726.1 | - | 4 | 635 | |||
BC069981.1 | - | 4 | 615 | 6336270 |