Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021472.4 | NP_067447.2 | 2 | 341 | ||
NM_201239.3 | NP_957691.1 | 2 | 638 | |||
GenBank mRNA | AB041044.1 | - | 2 | 247 | ||
AK050451.1 | - | 2 | 238 | |||
AY762362.1 | - | 2 | 240 | |||
BC005569.1 | - | 2 | 215 | 3967265 | ||
BC061105.1 | - | 2 | 222 | 30296907 |