Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278927.1 | NP_001265856.1 | 1, 2 | 271 | ||
NM_015342.3 | NP_056157.1 | 1, 2 | 274 | |||
XM_006714583.3 | XP_006714646.1 | 1, 2 | 167 | |||
XM_017009286.1 | XP_016864775.1 | 1, 2 | 46 | |||
GenBank mRNA | AK025679.1 | - | 1, 2 | 221 | ||
AK298175.1 | - | 1, 2 | 211 | |||
BC015385.1 | - | 1, 2 | 202 | 4390706 | ||
D38552.1 | - | 1, 2 | 197 | |||
DB454273.1 | - | 1, 2 | 271 |