Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289505.1 | NP_001276434.1 | 3 | 299 | ||
NM_021483.4 | NP_067458.2 | 3 | 431 | |||
GenBank mRNA | AB032591.1 | - | 3 | 426 | ||
AK039764.1 | - | 3 | 298 | |||
AK044552.1 | - | 2 | 451 | |||
BY129053.1 | - | 3 | 299 | |||
HQ646186.1 | - | 2 | 479 | |||
HQ646187.1 | - | 2 | 479 | |||
HQ646188.1 | - | 2 | 479 | |||
HQ646191.1 | - | 2 | 1135 | |||
HQ646192.1 | - | 3 | 549 | |||
HQ646193.1 | - | 3 | 455 | |||
HQ646194.1 | - | 3 | 455 | |||
HQ646195.1 | - | 3 | 323 | |||
HQ646196.1 | - | 3 | 323 |