Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163753.1 | NP_001157225.1 | 9 | 2274 | ||
NM_021411.4 | NP_067386.3 | 9 | 1886 | |||
GenBank mRNA | AK045995.1 | - | 9 | 1877 | ||
AK046260.1 | - | 9 | 2275 | |||
AK050483.1 | - | 9 | 1880 | |||
BC050757.1 | - | 9 | 1886 |