Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB041550.1 | - | 2 | 1353 | ||
BC139438.1 | - | 2 | 1347 | 8862460 | ||
BC139439.1 | - | 2 | 1347 | 8862461 |