Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021422.4 | NP_067397.1 | 5 | 919 | ||
GenBank mRNA | AB032401.1 | - | 6 | 786 | ||
AK003903.1 | - | 1 | 60 | |||
AK016666.1 | - | 4 | 516 | |||
AK046476.1 | - | 5 | 920 | |||
AK076175.1 | - | 5 | 920 | |||
AK141909.1 | - | 5 | 921 | |||
BC022948.1 | - | 3 | 417 | |||
BC147484.1 | - | 5 | 767 | 9056231 | ||
BC147486.1 | - | 5 | 767 | 9056239 |