Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021433.3 | NP_067408.1 | 4 | 507 | ||
GenBank mRNA | AF237814.1 | - | 4 | 317 | ||
AK009690.1 | - | 4 | 462 | |||
AK018167.1 | - | 4 | 495 | |||
AK019106.1 | - | 4 | 469 | |||
AK031673.1 | - | 4 | 486 | |||
AK044905.1 | - | 4 | 468 | |||
AK049124.1 | - | 4 | 467 | |||
AK151549.1 | - | 4 | 461 | |||
AK159392.1 | - | 4 | 479 | |||
AK179436.1 | - | 3 | 155 | |||
BC029205.1 | - | 4 | 424 | 4039560 | ||
BQ749205.1 | - | 4 | 480 |