Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253832.1 | NP_001240761.1 | 3, 4 | 502 | ||
NM_021438.4 | NP_067413.1 | 3, 4 | 502 | |||
GenBank mRNA | AF270700.1 | - | 3, 4 | 415 | ||
AK008093.1 | - | 3, 4 | 493 | |||
AK012629.1 | - | 3, 4 | 485 | |||
AK028029.1 | - | 3, 4 | 493 | |||
BC031716.1 | - | 3, 4 | 472 | 3969355 | ||
BC049284.1 | - | 3, 4 | 480 | 6395407 |