Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021447.2 | NP_067422.1 | 9 | 1512 | ||
GenBank mRNA | AF294790.1 | - | 9 | 1412 | ||
AK019655.1 | - | 5 | 468 |