Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163516.2 | NP_001156988.1 | 13 | 2060 | ||
NM_001163517.2 | NP_001156989.1 | 8 | 1182 | |||
NM_001289505.1 | NP_001276434.1 | 15 | 2041 | |||
NM_021483.4 | NP_067458.2 | 15 | 2170 | |||
GenBank mRNA | AB032591.1 | - | 15 | 2165 | ||
AK039764.1 | - | 15 | 2040 | |||
AK083141.1 | - | 13 | 1952 | |||
HQ646186.1 | - | 16 | 2389 | |||
HQ646187.1 | - | 15 | 2323 | |||
HQ646188.1 | - | 14 | 2218 | |||
HQ646189.1 | - | 14 | 2251 | |||
HQ646190.1 | - | 13 | 2146 | |||
HQ646191.1 | - | 15 | 2979 | |||
HQ646192.1 | - | 16 | 2393 | |||
HQ646193.1 | - | 16 | 2299 | |||
HQ646194.1 | - | 15 | 2194 | |||
HQ646195.1 | - | 16 | 2167 | |||
HQ646196.1 | - | 15 | 2062 | |||
HQ646197.1 | - | 8 | 1182 |