Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037085.1 | - | 7 | 629 | ||
BC049867.1 | - | 7 | 652 | |||
BC057921.1 | - | 7 | 640 | 5101105 | ||
X99571.1 | - | 7 | 635 |