Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190466.1 | NP_001177395.1 | 1 | 387 | ||
NM_021532.4 | NP_067507.2 | 1 | 387 | |||
GenBank mRNA | AF488775.1 | - | 1 | 287 | ||
AK198928.1 | - | 1 | 17 | |||
AY208970.1 | - | 1 | 239 | |||
BC145833.1 | - | 1 | 337 | 40131136 |