Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021546.3 | NP_067521.2 | 7 | 591 | ||
GenBank mRNA | AB039948.1 | - | 5 | 412 | ||
AK013520.1 | - | 7 | 590 | |||
AK044296.1 | - | 7 | 583 | |||
AK158225.1 | - | 7 | 593 | |||
BC126874.1 | - | 5 | 405 | 40044848 | ||
BC126875.1 | - | 5 | 405 | 40044849 |