Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105.4 | NP_001096.1 | 4 | 643 | ||
NM_001111067.2 | NP_001104537.1 | 4 | 462 | |||
XM_005246939.3 | XP_005246996.1 | 4 | 514 | |||
XM_005246940.3 | XP_005246997.1 | 4 | 482 | |||
XM_006712825.3 | XP_006712888.1 | 4 | 595 | |||
XM_011512106.2 | XP_011510408.1 | 5 | 562 | |||
XM_011512107.2 | XP_011510409.1 | 4 | 534 | |||
XM_011512108.2 | XP_011510410.1 | 4 | 1941 | |||
GenBank mRNA | AK297765.1 | - | 4 | 430 | ||
BC033867.1 | - | 4 | 643 | 5209711 | ||
BC036748.1 | - | 4 | 643 | |||
DA796090.1 | - | 4 | 462 | |||
KU177885.1 | - | 2 | 213 | |||
KU177886.1 | - | 2 | 213 | |||
KU177887.1 | - | 2 | 213 | |||
L02911.1 | - | 5 | 553 | |||
Z22534.1 | - | 3 | 316 |