Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005868.2 | NP_001005868.1 | 24 | 4368 | ||
NM_001289473.1 | NP_001276402.1 | 23 | 4224 | |||
NM_001289475.1 | NP_001276404.1 | 23 | 4251 | |||
GenBank mRNA | AF263743.1 | - | 3 | 1105 | ||
AK029054.1 | - | 13 | 3050 | |||
AK051733.1 | - | 3 | 883 | |||
AK122473.1 | - | 23 | 3825 | |||
BC005691.1 | - | 3 | 349 | |||
BC028256.1 | - | 4 | 1811 | |||
BC141188.1 | - | 24 | 4077 | 9055800 | ||
BC145498.1 | - | 23 | 3960 | 9054061 | ||
BC145509.1 | - | 23 | 3933 | 9054072 | ||
EU233994.1 | - | 26 | 5318 |