Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018727.5 | NP_061197.4 | 6 | 1147 | ||
NM_080704.3 | NP_542435.2 | 7 | 1264 | |||
NM_080705.3 | NP_542436.2 | 6 | 1190 | |||
NM_080706.3 | NP_542437.2 | 5 | 1516 | |||
GenBank mRNA | AF196175.1 | - | 6 | 1202 | ||
AF196176.1 | - | 6 | 1190 | |||
AJ272063.2 | - | 6 | 1147 | |||
AJ277028.1 | - | 5 | 1516 | |||
AL136801.1 | - | 7 | 1260 | |||
AY131289.1 | - | 5 | 989 | |||
AY986821.1 | - | 5 | 989 | |||
BC132820.1 | - | 5 | 1468 | 40146842 | ||
BC136633.1 | - | 5 | 1468 | 9020623 | ||
DQ177332.1 | - | 4 | 1076 | |||
DQ177333.1 | - | 5 | 1006 | |||
DQ898279.1 | - | 5 | 989 |