Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021568.2 | NP_067543.2 | 4, 5 | 596 | ||
GenBank mRNA | AF176327.1 | - | 4, 5 | 437 | ||
AK034811.1 | - | 7, 8 | 975 | |||
AK079564.1 | - | 2, 3 | 638 | |||
BC042440.1 | - | 2, 3 | 231 | 4511401 |