Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021790.2 | NP_068562.1 | 6, 7 | 688 | ||
NM_181061.5 | NP_851406.1 | 6, 7 | 811 | |||
GenBank mRNA | AB043687.1 | - | 6, 7 | 688 | ||
AK082939.1 | - | 6, 7 | 468 | |||
AK135773.1 | - | 204 | ||||
AK160776.1 | - | 6, 7 | 459 | |||
AK165268.1 | - | 1, 2 | 2895 | |||
BC028655.1 | - | 6, 7 | 824 |