Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018992.3 | NP_061865.1 | 2, 3 | 424 | ||
GenBank mRNA | AK000047.1 | - | 2, 3 | 422 | ||
BC007314.2 | - | 2, 3 | 377 | 2989642 | ||
DA134449.1 | - | 2, 3 | 424 |