Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021886.1 | NP_068686.1 | 1, 2 | 221 | ||
GenBank mRNA | AB017634.1 | - | 1, 2 | 221 | ||
AK006208.1 | - | 1, 2 | 206 | |||
AK011264.1 | - | 1, 2 | 206 | |||
AK011738.1 | - | 1, 2 | 206 | |||
BC025084.1 | - | 1, 2 | 172 | 3501244 |