Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021893.3 | NP_068693.1 | 3 | 166 | ||
GenBank mRNA | AF233517.1 | - | 2 | 99 | ||
AF317088.1 | - | 2 | 83 | |||
AK154403.1 | - | 3 | 145 | |||
BC066841.1 | - | 3 | 149 | 30101931 | ||
BY193530.1 | - | 3 | 162 | |||
GQ904196.1 | - | 2 | 83 |