Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022007.1 | NP_071290.1 | 6 | 640 | ||
GenBank mRNA | AK004113.1 | - | 6 | 649 | ||
BC048737.1 | - | 6 | 653 | |||
BC061101.1 | - | 6 | 644 | 6531288 |