Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021516.4 | NP_067491.2 | 6 | 1002 | ||
NM_022801.4 | NP_073712.2 | 6 | 1002 | |||
GenBank mRNA | AF240782.1 | - | 6 | 547 | ||
AF240783.1 | - | 6 | 547 | |||
AK083683.1 | - | 6 | 1003 | |||
AK132247.1 | - | 6 | 986 | |||
AK166157.1 | - | 6 | 991 | |||
AK220557.1 | - | 5 | 4836 | |||
BC066071.1 | - | 6 | 976 | |||
CX566304.1 | - | 3 | 140 |