Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190466.1 | NP_001177395.1 | 1, 2 | 425 | ||
NM_021532.4 | NP_067507.2 | 1, 2 | 425 | |||
GenBank mRNA | AF488775.1 | - | 1, 2 | 325 | ||
AK077691.1 | - | 1, 2 | 42 | |||
AK198928.1 | - | 1, 2 | 55 | |||
AY208970.1 | - | 1, 2 | 277 | |||
BC145833.1 | - | 1, 2 | 375 | 40131136 |