Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021554.2 | NP_067529.2 | 4 | 972 | ||
GenBank mRNA | AB041664.1 | - | 4 | 749 | ||
AJ278576.1 | - | 4 | 972 | |||
AK075586.1 | - | 4 | 735 | |||
AK169951.1 | - | 3 | 388 | |||
AK202674.1 | - | 2 | 182 | |||
AK206681.1 | - | 2 | 175 | |||
AK208061.1 | - | 2 | 182 | |||
AK209406.1 | - | 2 | 183 | |||
AK210173.1 | - | 2 | 182 | |||
AK216652.1 | - | 2 | 174 | |||
AK217664.1 | - | 2 | 182 | |||
AK219043.1 | - | 2 | 182 | |||
BC023188.1 | - | 3 | 413 | |||
BC068124.1 | - | 4 | 766 | 5716972 | ||
BC082317.1 | - | 3 | 397 |